
Chromium(III) potassium sulphate dodecahydrate ≥97%, GPR RECTAPUR®
Catalog # 21095.293
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 97 %
- Insolubility in water:Max. 0.2 %
- Cl (Chloride):Max. 0.02 %
- Cu (Copper):Max. 10 ppm
- Fe (Iron):Max. 0.02 %
- Pb (Lead):Max. 50 ppm
- Zn (Zinc):Max. 10 ppm
Specification Test Results
About this item
:
:499.4 g/mol
Merck Index:13,02250
Melting Pt:89 °C
Density:1.83 g/cm³ (20 °C)
EINECS:233-401-6
:GHS07
Storage Temperature:Ambient
CAS Number:7788-99-0
Formula:CrK(SO₄)₂·12H₂O
MDL Number:MFCD00149917