Magnesium acetate tetrahydrate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:16674-78-5
:
Formula:(H₃CCOO)₂Mg·4H₂O
MDL Number:MFCD00149214
:214.46 g/mol
Merck Index:13,05676
Melting Pt:80 °C
Density:1.454 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-554-9
Specification Test Results
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 100 ppm
- Ca (Calcium):Max. 0.02 %
- Fe (Iron):Max. 20 ppm
Specification Test Results
Frequently Bought Together