Order Entry
Slovenia
ContactUsLinkComponent
Dimethylammonium chloride, TECHNICAL
Irritant
Dimethylammonium chloride, TECHNICAL
Catalog # 23431.232
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Dimethylammonium chloride, TECHNICAL
Catalog # 23431.232
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :81.54 g/mol
Merck Index:13,03255
Melting Pt:170…173 °C
EINECS:208-046-5
 :GHS07
CAS Number:506-59-2
Formula:C₂H₈ClN
MDL Number:MFCD00012477