L(+)-Histidine monohydrochloride monohydrate
Supplier: Roth Carl
Certificates
About this item
Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate
CAS Number:5934-29-2
:
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151027
Density:1.485 g/cm³ (20 °C)
Molecular Weight:209.63 g/mol
Melting Pt:259
Merck Index:13,04741
Storage Temperature:Ambient
EINECS:211-438-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together