Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Plastic bottle, 4 L
Catalog # ROCH11666703001
Supplier: ROCHE
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # ROCH11666703001
Supplier: ROCHE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:4 L
- Description:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
- Cat. No.:ROCH11666703001
- M-Clarity quality level:MQ100



