Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Plastic bottle, 4 L
Catalog # ROCH11666690001
Supplier:   ROCHE
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Catalog # ROCH11666690001
Supplier:  ROCHE
CAS Number:  
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Plastic bottle, 4 L

CAS Number:  
Price
505,00 €
Log in for availability
1 * 4 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    4 L
  • Description:
    TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)
  • Cat. No.:
    ROCH11666690001
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.