Order Entry
Slovenia
ContactUsLinkComponent
Iodine 0.5 mol/l (1 N) in aqueous solution
Iodine 0.5 mol/l (1 N) in aqueous solution
Catalog # ROTHX868.1
Supplier:  Roth Carl
CAS Number:  
Iodine 0.5 mol/l (1 N) in aqueous solution
Catalog # ROTHX868.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
 :253.81 g/mol
Storage Temperature:Ambient
EINECS:231-442-4
UN:3264
ADR:8,II
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355