di-Sodium L(+)-tartrate dihydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6106-24-7
:
Formula:NaOOC(CHOH)₂COONa·2H₂O
MDL Number:MFCD00150035
:230.08 g/mol
Merck Index:14,8684
Melting Pt:∼150 °C (-2H₂O)
Density:1.82
Storage Temperature:Ambient
EINECS:212-773-3
Related Information
Frequently Bought Together