Polyethylene glycol 8,000, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779615
:8000 g/mol
Merck Index:12,07729
Boiling Pt:250 °C (1013 hPa)
Density:1.125 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-038-2
Frequently Bought Together