Order Entry
Slovenia
ContactUsLinkComponent
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Certificates
undefined
Polyethylene glycol 8,000
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White Flakes
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.5 % (K.F.)
  • Hydroxyl value:
    12 to 16 mg KOH/g