Order Entry
Slovenia
ContactUsLinkComponent
 
Glucose oxidase (from Aspergillus niger)
 :  Merck
 
Glucose oxidase (from Aspergillus niger)
 :  Merck
 

 

  • CAS No.:
    9001-37-0
  • Source:
    Aspergillus niger
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00131182
  • Enzyme name:
    Glucose oxidase

 

 

Frequently Bought Together