Starch (from corn), Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:9005-25-8
- Formula:(C₆H₁₀O₅)n
- MDL Number:MFCD00082026
- Molecular Weight:162.14 g/mol
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:232-679-6
2 Options Available Below
Product Details & Documents
- CAS Number:9005-25-8
- Formula:(C₆H₁₀O₅)n
- MDL Number:MFCD00082026
- Molecular Weight:162.14 g/mol
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:232-679-6
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



