Starch, soluble
Supplier: Roth Carl
Certificates
About this item
Synonyms: Starch, Vitex® iodine indicator, Amylodextrin
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
Molecular Weight:162.14 g/mol
Merck Index:13,08878
Storage Temperature:Ambient
EINECS:232-686-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together