Neomycin sulphate USP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1405-10-3
:MFCD00131817
Formula:C₂₃H₄₆N₆O₁₃·3H₂SO₄
MDL Number:MFCD00150827
Merck Index:12,06542
:908.87 g/mol
EINECS:215-773-1
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Potency (anhydrous):= 600 mcg/mg
- Loss on Drying:= 8 %
- pH (3.3%, Water) @25°C:5 - 7.5
Specification Test Results
Frequently Bought Together