D-(+)-Mannose, Sigma-Aldrich®
Catalog # SIAL63579-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:180.16 g/mol
Merck Index:13,05772
Boiling Pt:376 °C (1013 hPa)
Melting Pt:133 °C
Density:1.539 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:222-392-4
CAS Number:3458-28-4
:MFCD00799233
Formula:C₆H₁₂O₆
MDL Number:MFCD00064122