Rhodium, powder, Sigma-Aldrich®
Catalog # SIAL204218-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
ADR:4.1,III
:M-Clarity™ quality level = MQ100
:102.9 g/mol
EINECS:231-125-0
UN:3089
CAS Number:7440-16-6
Formula:Rh
MDL Number:MFCD00011201