Iridium ≥99.99% (metals basis), powder, Premion® -22 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-88-5
:
Formula:Ir
MDL Number:MFCD00011062
:192.22 g/mol
Merck Index:12,05098
Boiling Pt:4500 °C (1013 hPa)
Melting Pt:2443 °C
Density:22.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-095-9
UN:3089
ADR:4.1,III
Related Information
Frequently Bought Together