Tantalum, powder, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-25-7
:
Formula:Ta
MDL Number:MFCD00011252
:180.95 g/mol
Density:16.6 g/cm³ (20 °C)
EINECS:231-135-5
UN:3089
ADR:4.1,II
:M-Clarity™ quality level = MQ100
:GHS02
Frequently Bought Together