
Bismuth ≥99.999% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-69-9
:
Formula:Bi
MDL Number:MFCD00134033
:208.98 g/mol
Density:9.78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-177-4
UN:3089
ADR:4.1,III
:GHS02
Related Information
Frequently Bought Together