0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Indium
Indium ≥99.99% (metals basis), wire, Ø 1.5 mm (0.06 in)
Indium ≥99.99% (metals basis), wire, Ø 1.5 mm (0.06 in)
:
Thermo Fisher Scientific
Indium ≥99.99% (metals basis), wire, Ø 1.5 mm (0.06 in)
:
Thermo Fisher Scientific
:
7440-74-6
:
:
In
:
MFCD00134048
:
114.82 g/mol
:
13,04971
:
2000 °C (1013 hPa)
:
156 °C
:
7.3 g/cm³ (20 °C)
:
Ambient
:
231-180-0
Environmentally Preferable:
Documentation
SDS
msds_ALFA038517
Documentation
Frequently Bought Together
Water, HiPerSolv CHROMANORM®, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, TECHNICAL, demineralised
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals