Indium ≥99.998% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-74-6
:
Formula:In
MDL Number:MFCD00134048
Boiling Pt:2000
Melting Pt:156
Density:7.3 g/cm³ (20 °C)
Molecular Weight:114.82 g/mol
Merck Index:13,04971
Storage Temperature:Ambient
EINECS:231-180-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together