Order Entry
Slovenia
ContactUsLinkComponent
 
 
Magnesium 99.9+% (trace metals basis), turnings
 
undefined
 
Magnesium 99.9+% (trace metals basis), turnings

 

  • Appearance:
    Silver white Turnings
  • Assay:
    ≥99.90 %
  • Total trace metal impurities:
    ≤0.10 % (metals sought for: Si, Mn, Cu, Al, Fe, Pb, Ni, Sn)

 

 

Frequently Bought Together