Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Magnesium, granules

Magnesium, granules

Glass bottle for solids, 250 g
Catalog # SIAL63045-250G-F
Supplier:   Merck
Magnesium, granules
Catalog # SIAL63045-250G-F
Supplier:  Merck
CAS Number:  7439-95-4
Magnesium, granules

Glass bottle for solids, 250 g

CAS Number:  7439-95-4
Price
72,90 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:7439-95-4
  • Formula:Mg
  • MDL Number:MFCD00085308
  • Molecular Weight:24.3 g/mol
  • Merck Index:13,05675
  • Boiling Pt:1107 °C (1013 hPa)
  • Melting Pt:651 °C
  • Density:1.74 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-104-6
  • UN:1869
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications