

Poly(ethylene-co-acrylic acid), beads, Sigma-Aldrich®
SIAL426717-250G
: Merck
:
- Pack type:Poly bottle
- Pk:250 g
- Seq:3545
- Infrared Spectrum:Conforms to Structure
- Acid Value:37 - 44 mg/g
- Transition Temperature:99 - 101 °C
- Viscosity :575 cP (145 °C, Brookfield)(lit.)
- Drop Point:105 °C, Mettler
- Hardness:≤2.5 dmm (ASTM D 5, 25 °C)
- Composition :Acrylic Acid, 5 wt.%
:
:385.371 g/mol
:0.93 g/ml (25 °C)
:8,I
:M-Clarity™ quality level = MQ100
:3261
:GHS05
:GHS07
:9010-77-9
:C₁₉H₁₉N₃O₆
:MFCD00133995