Order Entry
Slovenia
ContactUsLinkComponent
L-key set, compact holder
L-key set, compact holder
Catalog # 238-1101
L-key set, compact holder
Catalog # 238-1101

Specifications

  • Description:
    L-key set in compact holder
  • Pk:
    1 SET

Specifications

About this item

These matt chrome-plated keys have a hexagonal ball end.

  • Fits easily into every ruler pocket
  • Especially suitable for screws that are difficult to access
  • Ball end enables the user to work at angles up to 25°
Certifications: Based on ISO 2936L.
Delivery: The set includes 9 pieces.
Packaging: Packed in blister pack.