Human recombinant PI3 kinase p110 alpha/p85 alpha
Catalog # ENZOBMLSE4360020
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:20 µG
- Enzyme type:Recombinant
- Source:
- Species:Human
- Seq:02
- Environmentally Preferable:
- Enzyme name:PI3 kinase p110 alpha/p85 alpha
Specifications
About this item
Produced in insect cells. Complex of human full length p110a and p85a subunits of phosphatidylinositol 3-kinase.