Human recombinant MMP7 (catalytic domain) (from E. coli)
ENZOBMLSE1810010
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:MMP7 (catalytic domain)
Produced in E. coli. Active Matrix Metalloproteinase-7 (MMP-7, matrilysin, pump) catalytic domain from human cDNA.
The enzyme consists of the catalytic domain of human MMP-7 (Tyr95-Lys267, NM_002423) with a C-terminal purification tag.