Order Entry
Slovenia
ContactUsLinkComponent
pp60v-src autophosphorylation site
pp60v-src autophosphorylation site
Catalog # ENZOBMLP3070001
Supplier:  Enzo Life Sciences
CAS Number:  
pp60v-src autophosphorylation site
Catalog # ENZOBMLP3070001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    pp60v-src autophosphorylation site
  • Purity:
    97%

Specifications

About this item

EGFR substrate

Residues 2-12 are the v-Src (412-422) autophosphorylation site with an extra Arg for phosphocellulose binding. This substrate sequence, RRLIEDNEYTARG, was originally described as an EGFR substrate in A431 cell extracts (Km=0,3-0,6 mM).