CDK2 substrate
ENZOBMLP2650001
New Product
:
- Pk:1 mg
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:CDK2 Substrate
- Purity:>95% (HPLC)
- Formulation:Lyophilised
This peptide, HHASPRK, is a specific substrate for CDK2.