ApoE ELISA kit, grow cells
Catalog # ENZOENZKIT1340001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Description:ApoE ELISA kit, grow cells
- Seq:01
- Sensitivity:14,063 ng/ml (23,438 - 1500 ng/ml)
Specifications
About this item
Highly sensitive ApoE ELISA kit enabling detection of ApoE in serum, plasma, cell culture supernates, and tissue homogenates in 3,5 hours.
- Sensitive measurement of ApoE, detecting as little as 14,063 pg/ml
- Negligible reactivity to similar proteins.
- High throughput with results in 3,5 hours.
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ApoE in serum, plasma, cell culture supernates and tissue homogenates in human samples.
The ApoE ELISA kit is a colorimetric immunometric immunoassay kit with results in 3,5 hrs.
Ordering information: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Conjugate Buffer Substrate, and Stop Solution.