Gastrin I (rat), ELISA kit
ENZOADI900149
:
- Pk:1 KIT
- Assay Type:Competitive
- Primary antibody reactivity:Rat
- Target protein:Gastrin I
- Description:Gastrin I (rat), ELISA kit
- Seq:02
- Sensitivity:78,1 pg/ml (range 78,1 - 5,000 pg/ml)
For the quantitative determination of rat Gastrin I in culture supernatants, plasma, and serum.
The Gastrin I (rat), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error in the lab. This kit utilizes low sample volume and the ability to measure up to 37 samples in duplicate per 96-well plate.
: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.