Order Entry
Slovenia
ContactUsLinkComponent
LBP, soluble (mouse) ELISA kit
LBP, soluble (mouse) ELISA kit
Catalog # ENZOALX850305KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
LBP, soluble (mouse) ELISA kit
Catalog # ENZOALX850305KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    LBP
  • Description:
    LBP, soluble (mouse) ELISA kit
  • Seq:
    02
  • Sensitivity:
    Range: 1 - 50 ng/ml

Specifications

About this item

For the quantitative determination of mouse and rat soluble as well as bound LBP levels in serum, plasma and cell culture medium.

Ordering information: Contains 1 precoated ELISA module1 vial anti-LBP (mouse) detecting MAb1 vial LBP standard (mouse)1 vial reference serum (mouse)2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution1 vial substrate solution.