CD40L Soluble (Human) ELISA kit
Catalog # ENZOALX850311KI01
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:CD40
- Description:CD40L soluble (Human) ELISA kit
- Seq:02
- Storage conditions:4 °C (kit), −20 °C (control)
- Sensitivity:0,005 ng/ml (Range 0,08 - 5 ng/ml)
Specifications
About this item
For the quantitative determination of soluble human CD40L (CD154) in cell culture supernatants, serum and other body fluids.
- Assay Time: 3 hours + 10 minutes