Testosterone high sensitivity ELISA kit
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Testosterone ELISA kit is a highly sensitive competitive immunoassay for the measurement of testosterone in human serum and plasma samples. Results are read on a colorimetric plate reader at 405 nm.
- Excellent sensitivity
- Validated for use with human samples
- Stable, colour coded reagents reduce errors
Delivery: Kit includes GxM microtiter plate, assay buffer, standard, antibody, conjugate, wash buffer concentrate, pNpp substrate, stop solution.
Specifications
- Assay Type:Competitive
- Primary antibody reactivity:Human
- Target protein:Testosterone
- Storage conditions:4 °C (short and long term)
- Sensitivity:2,6 pg/ml (range 3,9 - 1000 pg/ml)
Specifications
Frequently Bought Together