Big endothelin-1 (rat), ELISA kit
Catalog # ENZOADI900073
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Rat
- Target protein:Endothelin-1
- Description:Big Endothelin-1 (Rat), ELISA kit
- Seq:02
- Storage conditions:4 °C (long term)
- Sensitivity:0,72 pg/ml (range 0,78 - 100 pg/ml)
Specifications
About this item
The Big Endothelin-1 (Rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results overnight + 1 hour.
- Measure low levels of rat big endothelin-1
- Convenient method
- Assay up to 38 samples in duplicate per plate
For the quantitative determination of rat Big ET-1 in culture supernatants, plasma, and serum.
Delivery: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.