About this item
The Hsp70 family of heat shock proteins contains multiple homologs ranging in size from 66-78 kDa, and are the eukaryotic equivalents of the bacterial DnaK. The most studied Hsp70 members include the cytosolic stress-induced Hsp70 (Hsp72), the constitutive cytosolic Hsc70 (Hsp73), and the ER-localized BiP (Grp78). Hsp70 family members contain highly conserved N-terminal ATP-ase and C-terminal protein binding domains. Binding of peptide to Hsp70 is assisted by Hsp40, and stimulates the inherent ATPase activity of Hsp70, facilitating ATP hydrolysis and enhanced peptide binding. Hsp70 nucleotide exchange and substrate binding coordinates the folding of newly synthesized proteins, the re-folding of misfolded or denatured proteins, coordinates trafficking of proteins across cellular membranes, inhibits protein aggregation, and targets the degradation of proteins via the proteasomal pathway.
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: C92F3A-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Fish, Drosophila, Dog, Chicken, C. elegans, Bovine
Specifications
- Catalog No:
- ENZOADISPA810488F
- ENZOADISPA810488D
- Antigen symbol:
- HSP70
- HSP70
- Antigen name:
- Heat Shock Protein 70
- Heat Shock Protein 70
- Antigen synonyms:
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- DyLight® 488
- Clone:
- C92F3A-5
- C92F3A-5
- Reactivity:
- Fish, Drosophila, Chicken, C. elegans, Human, Bovine, Dog, Rat, Mouse
- Fish, Drosophila, Chicken, C. elegans, Human, Bovine, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Native human Hsp70
- Native human Hsp70
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein G affinity chromatography
- Purified by Protein G affinity chromatography
- Storage buffer:
- PBS containing 0,09% sodium azide
- PBS containing 0,09% sodium azide
- Storage temperature:
- 4 °C
- 4 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice