Anti-CGMP Rabbit Polyclonal Antibody
ENZOADI915063100
- Antibody type:Primary
- Antigen name:Cyclic guanosine monophosphate
- Antigen symbol:cGMP
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Multiple
- Seq:02
- Form:Liquid. Contains 0,1% sodium azide.
- Antigen synonyms:Cyclic GMP
- Pk:100 µl
Cyclic GMP (cGMP), which has been shown to be present at levels typically 10-100 fold lower than cAMP, is formed by the action of the enzyme guanylate cyclase on GTP. Some hormones, such as Acetylcholine, Insulin, and Oxytocin, as well as certain other chemicals like Serotonin and Histamine cause an increase in cGMP levels. Stimulators of guanylate cyclase such as the vasodilators nitroprusside, nitroglycerin, sodium nitrate, and nitric oxide (NO) also stimulate cGMP levels.
- For ELISA, Flow Cytometry
Species independent.
Type: Primary
Antigen: CGMP
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: