
3-Sulphopropyl methacrylate potassium salt 96.0-104.0% (by titrimetric analysis), Sigma-Aldrich®
Catalog # SIAL251631-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:246.32 g/mol
Melting Pt:302 °C (lit.)
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:31098-21-2
Formula:C₇H₁₂O₅S
MDL Number:MFCD00010139