Order Entry
Slovenia
ContactUsLinkComponent
 
 
Disodium succinate, anhydrous for biochemistry
 
undefined
 
Disodium succinate, anhydrous for biochemistry

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.3 %
  • UV:
    (0.1 M)
  • pH:
    6 to 9 (0.4 mol sol)
  • Titration with HClO4:
    98.5 to 102 %
  • Heavy metals (as Pb):
    ≤10 ppm

 

Frequently Bought Together