
Primaquine diphosphate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63-45-6
:
Formula:C₁₅H₂₇N₃O₉P₂
MDL Number:MFCD00013166
:455.33 g/mol
Melting Pt:205…206 °C (dec.)
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Orange to red-orange Powder
- Infrared spectrum:Conforms
- Melting point:199 to 205 °C
- Titration with NaOH:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together