Push caps, TPE, Capcluster-96
Supplier: MICRONIC
Certificates
About this item
Capcluster-96 TPE push caps are designed to individually seal Micronic tubes in 96-well format.
- Multi-pierceable without losing seal integrity
- Choice of colours to aid identification
- Class 7 cleanroom production
- Free from RNase, DNase and pyrogens
- Resistant to chemical solvents
Frequently Bought Together