Dental micro spatulas, double-ended, Aesculap®
Supplier: Aesculap
Certificates
About this item
Micro spatulas for application of the retrograde filling material (Super EBA) into the dental cavity.
Frequently Bought Together
Micro spatulas for application of the retrograde filling material (Super EBA) into the dental cavity.