Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Gas distribution tubes

Gas distribution tubes

4, 250 mm, 6 mm, 20 mm
Catalog # 820-3050
Gas distribution tubes
Catalog # 820-3050
Gas distribution tubes
Gas distribution tubes

4, 250 mm, 6 mm, 20 mm

Price
36,05 €
Log in for availability
1 * 1 Each
Product Details & Documents

These gas distribution tubes have a laterally fused-on bowl with filter disc.

Specifications
  • Head Ø:
    20 mm
  • Porosity:
    4
  • Tube ext.Ø:
    6 mm
  • Length:
    250 mm