Order Entry
Slovenia
ContactUsLinkComponent
Econazole Related Compounds, Reference Standard
Econazole Related Compounds, Reference Standard
Supplier:  USP
Certificates
Econazole Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    24169-02-6
  • Formula:
    C₁₈H₁₆Cl₃N₃O₄
  • Description:
    Econazole Nitrate
  • Synonym(s):
    2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine nitrate

Specifications

Related Information

Frequently Bought Together