Order Entry
Slovenia
ContactUsLinkComponent
Doxycycline Related Compounds, Reference Standard
Doxycycline Related Compounds, Reference Standard
Supplier:  USP
Certificates
Doxycycline Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    24390-14-5
  • Formula:
    C₄₆H₅₈Cl₂N₄O₁₈
  • Description:
    Doxycycline Hyclate
  • Synonym(s):
    (4S,4aR,5S,5aR,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide

Specifications

Frequently Bought Together