Order Entry
Slovenia
ContactUsLinkComponent
Diclofenac Related Compounds, Reference Standard
Diclofenac Related Compounds, Reference Standard
Supplier:  USP
Certificates
Diclofenac Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    15307-79-6
  • UN No.:
    2811
  • Description:
    Diclofenac Sodium
  • Formula:
    C₁₄H₁₀Cl₂KNO₂
  • Synonym(s):
    N-(2,6-dichlorophenyl)indolin-2-one

Specifications

Related Information

Frequently Bought Together