Order Entry
Slovenia
ContactUsLinkComponent
Desoxycorticosterone Related Compounds, Reference Standard
Desoxycorticosterone Related Compounds, Reference Standard
Supplier:  USP
Certificates
Desoxycorticosterone Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    56-47-3
  • Formula:
    C₂₃H₃₂O₄
  • Description:
    Desoxycorticosterone Acetate

Specifications

Frequently Bought Together