Order Entry
Slovenia
ContactUsLinkComponent
Clomiphene Related Compounds, Reference Standard
Clomiphene Related Compounds, Reference Standard
Supplier:  USP
New Product
Certificates
Clomiphene Related Compounds, Reference Standard
Supplier:  USP
New Product

Specifications

  • CAS No.:
    50-41-9
  • Formula:
    C₃₂H₃₆ClNO₈
  • Description:
    Clomiphene citrate
  • Synonym(s):
    (E,Z)-2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride (1:1)

Specifications

Frequently Bought Together