Order Entry
Slovenia
ContactUsLinkComponent
Human recombinant Death Receptor 6 (from cells)
Human recombinant Death Receptor 6 (from cells)
Catalog # PRSI92-146
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Death Receptor 6 (from cells)
Catalog # PRSI92-146
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    TNFRSF21|UNQ437/PRO868|DR6|CD358|Death Receptor 6|Tumor Necrosis Factor Receptor Superfamily Member 21
  • Protein/peptide name:
    Death Receptor 6
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    61.7 kD
  • Sequence:
    Gln42-Leu350
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Tumour Necrosis Factor Receptor Superfamily Member 21 (TNFRSF21) is a type I transmembrane receptor that includes four extracellular cysteine-rich motifs and a cytoplasmic death domain. DR6 is highly expressed in heart, brain, placenta, pancreas, lymph node, thymus and prostate. DR6 may activate NF-kappa-B and JNK to promote apoptosis and T-cell differentiation. In addition, DR6 binds with N-APP, which is released by the deprivation of Trophic-factor. It triggers caspase activation and degeneration of both neuronal cell bodies (via caspase-3) and axons (via caspase-6). DR6 is also expressed on the tumour cell lines and can be induced by TNF- alpha.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.