Human recombinant SOD2 (from E. coli)
Catalog # PRSI91-566
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:SOD2|Mn-SOD|Superoxide Dismutase [Mn] Mitochondrial
- Protein/peptide name:SOD2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:23.24 kD
- Sequence:Met1-Lys222
- Formulation:Supplied as a 0.2 um filtered solution of 20mM Tris,100mM NaCl,50% glycerol,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Superoxide Dismutase (SOD2) is a number of the iron/manganese superoxide dismutase family. SOD2 is a mitochondrial protein that forms a homotetramer and binds one manganese ion per subunit. The SOD2 protein transforms toxic superoxide and a byproduct of the mitochondrial electron transport chain into hydrogen peroxide and diatomic oxygen. Genetic variation in SOD2 is associated with microvascular complications of diabetes type 6 (MVCD6), idiopathic cardiomyopathy (IDC), sporadic motor neuron disease, and cancer. SOD2 destroys superoxide anion radicals which are usually produced within the cells and which are toxic to biological systems.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag